Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAB2IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585697 Supplier Invitrogen™ Supplier No. PA585697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (98%), Bovine (98%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAB2IP
Applications Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Dab2ip
Gene Accession No. Q5VWQ8
Gene Alias 2310011D08Rik; AF9Q34; AI480459; AIP1; AIP-1; ASK1-interacting protein 1; ASK-interacting protein 1; DAB2 interacting protein; DAB2 interaction protein; DAB2-interacting protein; Dab2ip; DIP1/2; disabled 2 interacting protein; disabled 2 interacting protein long form; disabled homolog 2 interacting protein; disabled homolog 2-interacting protein; DOC2/DAB2 interactive protein; DOC-2/DAB2 interactive protein; DOC-2/DAB-2 interactive protein; Kiaa1743; mKIAA1743; nGAP-like protein
Gene Symbols Dab2ip
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human DAB2IP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153090
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.