Learn More
DAB2IP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302439AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.Specifications
DAB2IP | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q3UHC7, Q5VWQ8 | |
DAB2IP | |
Between 850 and 900 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DAB2IP | |
Disabled homolog 2-interacting protein, DAB2 interaction protein, DAB2-interacting protein, ASK-interacting protein 1, AIP-1, DOC-2/DAB-2 interactive protein, DAB2IP, AF9Q34, AIP1, KIAA1743 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
153090, 69601 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.