Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAB2IP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986963
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6963 Supplier Bioss Supplier No. BS5998R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.

Specifications

Antigen DAB2IP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Dab2ip
Gene Accession No. Q5VWQ8
Gene Alias 2310011D08Rik; AF9Q34; AI480459; AIP1; AIP-1; ASK1-interacting protein 1; ASK-interacting protein 1; DAB2 interacting protein; DAB2 interaction protein; DAB2-interacting protein; DAB2IP; DIP1/2; disabled 2 interacting protein; disabled 2 interacting protein long form; disabled homolog 2 interacting protein; Disabled homolog 2-interacting protein; DOC2/DAB2 interactive protein; DOC-2/DAB2 interactive protein; DOC-2/DAB-2 interactive protein; KIAA1743; mKIAA1743; nGAP-like protein
Gene Symbols Dab2ip
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DAB2IP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153090
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.