Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dabsyl-L-methionine 95.0+%, TCI America™
SDP

Catalog No. d1457100mg Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 97684-99-6
Molecular Formula C19H24N4O4S2
Molecular Weight (g/mol) 436.545
MDL Number MFCD00059323
InChI Key YGFUZBPDTRWEQW-SFHVURJKSA-N
Synonym 4-Dimethylaminoazobenzene-4′C-sulfonyl-L-methionine, Dbs-Met-OH
PubChem CID 14048711
IUPAC Name (2S)-2-[[4-[[4-(dimethylamino)phenyl]diazenyl]phenyl]sulfonylamino]-4-methylsulfanylbutanoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)NC(CCSC)C(=O)O

Specifications

Quantity 100 mg
Formula Weight 436.55
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dabsyl-L-methionine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.