Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dacarbazine, Fisher BioReagents

Catalog No. p-25356 Shop All Fisherbrand Products
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass
Dacarbazine is an alkylating agent used in cancer research. Antineoplastic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4342-03-4
Molecular Formula C6H10N6O
Molecular Weight (g/mol) 182.19
InChI Key OMJKFYKNWZZKTK-UXBLZVDNSA-N
Synonym dacarbazine, biocarbazine, deticene, dtic-dome, biocarbazine r, decarbazine, dtic, icdt, dacarbazinum inn-latin, dacarbazino inn-spanish
PubChem CID 5353562
IUPAC Name (5E)-5-(dimethylaminohydrazinylidene)imidazole-4-carboxamide
SMILES CN(C)NN=C1C(=NC=N1)C(=O)N

Specifications

Melting Point 202°C
Color White
Packaging Amber Glass
Quantity 5 g
Merck Index 15, 2795
Formula Weight 182.2
Physical Form Powder or Solid
Chemical Name or Material Dacarbazine
Recommended Storage 4° C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.