Learn More
Invitrogen™ DACH1 Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 83% homologous in Zebrafish.
Specifications
Specifications
| Antigen | DACH1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | Dach1 |
| Gene Accession No. | Q9QYB2, Q9UI36 |
| Gene Alias | Dac; dac homolog; DACH; Dach1; dachshund 1; dachshund 1 (Drosophila); dachshund family transcription factor 1; dachshund homolog 1; FLJ10138; OTTHUMP00000040839; RP11-512J14.1 |
| Gene Symbols | Dach1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.