Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DACH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551369 Supplier Invitrogen™ Supplier No. PA551369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITAAAMAEAM KLQKMKLMAM NTLQGNGSQN GTESEPDDLN SNTGGSESSW DKDKMQSPFA APGPQHGIAH AALAGQPGIG GAPTLNPLQQ NHLLTNRLDL PFMMMPHPLL PVSLPPASV Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 59%.

This gene is one of two genes which encode a protein similar to the Drosophila protein dachshund, a transcription factor involved in cell fate determination in the eye, limb and genital disc of the fly. The encoded protein contains two characteristic dachshund domains: an N-terminal domain responsible for DNA binding and a C-terminal domain responsible for protein-protein interactions. This gene is located on the X chromosome and is subject to inactivation by DNA methylation. The encoded protein may be involved in regulation of organogenesis and myogenesis, and may play a role in premature ovarian failure. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DACH2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dach2
Gene Accession No. Q96NX9
Gene Alias 9430028N04Rik; DACH2; dachshund 2; dachshund 2 (Drosophila); dachshund family transcription factor 2; dachshund homolog 2; FLJ31391; MGC138545; RGD1561687; RP11-345E19.1
Gene Symbols Dach2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DACH2. Recombinant protein control fragment (Product #RP-92001).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.