Learn More
DACT1 Mouse anti-Human, Clone: OTI10F12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809184

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Dapper homolog 1, an ortholog of Xenopus Dpr, antagonizes Wnt signaling by promoting Dvl degradation via a lysosome inhibitor-sensitive and proteasome inhibitor-insensitive mechanism. Dapper homolog 1 acts as an interacting protein for Dvl, and the interaction is formed between the DEP (Dishevelled, Egl-10, and pleckstrin) domain of Dvl and the central and C-terminal regions of Dapper homolog 1. The gene for human Dapper homolog 1 is localized on chromosomal region 14q23.1. The protein impedes the degradation of CTNNB1/beta-catenin, thereby enhancing the transcriptional activation of target genes of the Wnt signaling pathway. The protein is also required for normal notochord formation and is downregulated in hepatocellular carcinomas (HCC). It is expressed in human liver and various other tissues.Specifications
DACT1 | |
Monoclonal | |
Unconjugated | |
DACT1 | |
DAPPER, DAPPER1, DPR1, FRODO, HDPR1, TBS2, THYEX3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51339 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI10F12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NYF0 | |
DACT1 | |
Human recombit protein fragment corresponding to amino acids 289-555 of human DACT1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.