Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DACT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551796 Supplier Invitrogen™ Supplier No. PA551796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPLPYASPYA YVASDSEYSA ECESLFHSTV VDTSEDEQSN YTTNCFGDSE SSVSEGEFVG ESTTTSDSEE SGGLIWSQFV QTLPIQTVTA PDLHNHPAKT FVKIKASHNL KKKILRFRSG S Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

Dapper homolog 1, an ortholog of Xenopus Dpr, antagonizes Wnt signaling by promoting Dvl degradation via a lysosome inhibitor-sensitive and proteasome inhibitor-insensitive mechanism. Dapper homolog 1 acts as an interacting protein for Dvl, and the interaction is formed between the DEP (Dishevelled, Egl-10, and pleckstrin) domain of Dvl and the central and C-terminal regions of Dapper homolog 1. The gene for human Dapper homolog 1 is localized on chromosomal region 14q23.1. The protein impedes the degradation of CTNNB1/beta-catenin, thereby enhancing the transcriptional activation of target genes of the Wnt signaling pathway. The protein is also required for normal notochord formation and is downregulated in hepatocellular carcinomas (HCC). It is expressed in human liver and various other tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DACT1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DACT1
Gene Accession No. Q9NYF0
Gene Alias 4921528D17Rik; AI115603; dact1; dact1.S; dact1-a; dact1-b; DAPPER; dapper 1; dapper 1-A; dapper antagonist of catenin 1; Dapper homolog 1; dapper homolog 1, antagonist of beta-catenin; dapper homolog 1, antagonist of beta-catenin (xenopus); dapper, antagonist of beta-catenin; dapper, antagonist of beta-catenin, homolog 1; dapper1; dapper1b; dishevelled binding antagonist of beta catenin 1; dishevelled-binding antagonist of beta-catenin 1; dishevelled-binding antagonist of beta-catenin 1 S homeolog; dishevelled-interacting protein; dpr; dpr1; frd1; FRODO; frodo 1; frodo homolog; Frodo1; functional regulator of dsh in ontogenesis; HDPR1; Hepatocellular carcinoma novel gene 3 protein; heptacellular carcinoma novel gene 3; HNG3; MDpr1; MTNG3; RGD1564008; signaling protein; thyex3; thymus expressed gene 3; thymus-expressed novel gene 3 protein; XDpr; XDpr1b; XELAEV_18041122mg
Gene Symbols DACT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DACT1. Recombinant protein control fragment (Product #RP-88940).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51339
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.