Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DACT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556415
Description
Immunogen sequence: PWGTFWPRPV STGDLDRALP ADTGLQKASA DAELLGLLCQ GVDIPLHVPD PKYRQDLVSQ GGREVYPYPS PLHAVALQSP LFVLTKETPQ Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 70%.
The Wnt signaling cascade is a conserved process in multicellular animals that plays important roles during development and can contribute to cancer and other diseases. Many members of this pathway are also expressed in the postnatal tissues such as brain. One such protein is Dact2, a member of the Dact protein family that was initially identified through binding to Disheveled (Dvl), a cytoplasmic protein essential to Wnt signaling. Dact2 is most prominent during the development of the thymus kidneys, and salivary gland. Dact2 is thought to play a role distinct from that of Dact1 with Dact2 having a greater impact on a beta-catenin-independent process termed planar cell polarity/convergent-extension signaling. Furthermore, Dact2 but not Dact1 can inhibit Nodal signaling by promoting the endocytic degradation of TGF-beta receptors. At least two isoforms of Dact2 are known to exist.
Specifications
DACT2 | |
Polyclonal | |
Unconjugated | |
DACT2 | |
2900084M21Rik; A630024E20; bA503C24.7; C6orf116; Dact2; Dapper antagonist of catenin 2; dapper homolog 2; dapper homolog 2, antagonist of beta-catenin; dapper homolog 2, antagonist of beta-catenin (xenopus); dapper, antagonist of beta-catenin, homolog 2; DAPPER2; dishevelled binding antagonist of beta catenin 2; dishevelled-binding antagonist of beta-catenin 2; Dpr2; Frd2; mDpr2; PP13671; RP11-503C24.7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
168002 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5SW24 | |
DACT2 | |
Recombinant protein corresponding to Human DACT2. Recombinant protein control fragment (Product #RP-94929). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction