Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DACT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560217 Supplier Invitrogen™ Supplier No. PA560217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALEEQLEALP GLVWDLGQQL GDLSLESGGL EQESGRSSGF YEDPSSTGGP DSPPSTFCGD SGFSGSSSYG RLGPSEP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The Wnt signaling cascade is a conserved process in multicellular animals that plays important roles during development and can contribute to cancer and other diseases. Many members of this pathway are also expressed in the postnatal tissues such as brain. One such protein is Dact3, a member of the Dact protein family that was initially identified through binding to Disheveled (Dvl), a cytoplasmic protein essential to Wnt signaling. Dact3 is expressed in the ventral region of maturing somites, limb bud and branchial arch mesenchyme, embryonic CNS, and the adult brain. Recent evidence shows that Dact3 acts as a negative regulator Wnt/beta-catenin signaling that is repressed at the transcriptional level in colorectal cancer and this repression is associated with bivalent histone modifications. This repression can be reversed by pharmacological agents that targets both histone methylation and deacetylation, suggesting that Dact3 may be a potential target for therapeutic treatment of this cancer. At least three isoforms of Dact3 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DACT3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DACT3
Gene Accession No. Q96B18
Gene Alias Antagonist of beta-catenin Dapper homolog 3; arginine-rich region 1 protein; dact 3; Dact3; Dapper antagonist of catenin 3; dapper homolog 3; dapper homolog 3, antagonist of beta-catenin (xenopus); dapper, antagonist of beta-catenin, homolog 3; DAPPER3; dishevelled binding antagonist of beta catenin 3; dishevelled-binding antagonist of beta-catenin 3; EG666671; naa-3; RGD1563892; RRR1
Gene Symbols DACT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DACT3. Recombinant protein control fragment (Product #RP-99202).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.