Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Dade Actin™ Activated Cephaloplastin Reagent

Catalog No. p-7235886 Shop All Siemens Healthineers Products
Click to view available options
Quantity:
10 x 2 mL
10 x 10 mL
This item is not returnable. View return policy

Catalog No. 23-044-646

Supplier: Siemens Healthineers 10445709

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ideal choice for institutions requiring a moderate screening APTT reagent for routine testing.

Dade Actin™ Activated Cephaloplastin reagent has moderate sensitivity to factor deficiencies (VIII, IX, XI, and XII) in the intrinsic system. It is the ideal choice for institutions requiring a moderate screening APTT reagent for routine testing. Dade Actin™ Activated Cephaloplastin reagent has low heparin sensitivity, allowing the monitoring of heparin therapy even with high heparin dosage. It has moderate sensitivity to lupus anticoagulants.
TRUSTED_SUSTAINABILITY

Specifications

Type Hemostasis Reagent
Quantity 10 x 2 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.