Learn More
Siemens Healthineers Dade Actin™ FS Activated PTT Reagent
Catalog No. p-7235894 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 2 mL
10 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-649 10 x 2 mL
23-044-647 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-649 Supplier Siemens Healthineers Supplier No. 10445712
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fulfills all requirements of routine coagulation testing.
Dade Actin™ FS Activated PTT reagent is a highly sensitive reagent for the detection of factor deficiencies (VIII, IX, XI and XII) of the intrinsic system. With moderate sensitivity to lupus anticoagulants and high sensitivity to heparin, it fulfills all requirements of routine coagulation testing.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
Quantity 10 x 2 mL