Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Dade Actin™ FSL Activated PTT Reagent

Catalog No. p-7235907 Shop All Siemens Healthineers Products
Click to view available options
Quantity:
10 x 2 mL
10 x 10 mL
This item is not returnable. View return policy

Catalog No. 23-044-650

Supplier: Siemens Healthineers 10445713

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Shows good factor sensitivity to detect clinically significant deficiencies of the intrinsic system.

Dade Actin™ FSL Activated PTT reagent exhibits increased sensitivity to lupus anticoagulants and moderate heparin sensitivity. The reagent shows good factor sensitivity to detect clinically significant deficiencies of the intrinsic system.
TRUSTED_SUSTAINABILITY

Specifications

Type Hemostasis Reagent
Quantity 10 x 2 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.