Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Dade™ Innovin™ PT Reagent (US)

Catalog No. p-7235905 Shop All Siemens Healthineers Products
Click to view available options
Quantity:
10 x 4 mL
10 x 10 mL
12 x 20 mL
This item is not returnable. View return policy

Catalog No. 23-044-776

Supplier: Siemens Healthineers 10873566

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use in prothrombin time (PT) determinations and prothrombin time-based assays in citrated human plasma.

Dade™ Innovin™ Reagent is prepared from purified recombinant human tissue factor produced in E. coli combined with synthetic phospholipids, calcium, buffers, and stabilizers. The reagent contains minimal residual clotting factors, such as prothrombin and factors VII or X, for clear definition of factor deficiencies and steep factor assay curves. It is highly sensitive to extrinsic factor deficiencies and oral anticoagulant-treated patient plasma samples.

The sensitivity of the Dade™ Innovin™ Reagent is very similar to the WHO human brain reference thromboplastin. It is insensitive to therapeutic levels of heparin, which, in combination with its high sensitivity to coagulation factors, makes Dade™ Innovin™ ideal for monitoring oral anticoagulant therapy and differentiating abnormal plasmas, even in the mildly pathological range.

TRUSTED_SUSTAINABILITY

Specifications

Type Hemostasis Reagent
For Use With (Application) Prothrombin Time (PT) Testing
Quantity 10 x 4 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.