Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAF-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-166 Supplier Invitrogen™ Supplier No. PA116925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAF-3 Polyclonal Antibody for Western Blot, IHC (P), IP, ChIP

Signals from TGF-beta superfamily receptors are transduced to the nucleus by Smad proteins, which transcriptionally activate target genes. In Caenorhabditis elegans (C.elegans), defects in a TGF-beta-related pathway cause a reversible developmental arrest and metabolic shift at the dauer larval stage. DAF-3 transgenes interfere with dauer formation, playing a role in the nucleus. DAF-3 is unique in that it is antagonized, rather than activated, by a TGF-beta pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAF-3
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene daf-3
Gene Accession No. Q95QI7
Gene Alias CELE_F25E2.5; daf-3; Dwarfin sma;MH2 domain-containing protein; protein-coding; Smad protein
Gene Symbols daf-3
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus (within residues 700-796) of the C.elegans DAF-3 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 180431
Target Species C. elegans
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.