Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAG Kinase Alpha Mouse anti-Human, Clone: 10D6.1, MilliporeSigma™

Catalog No. MABF1105MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1105MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1105MI Supplier MilliporeSigma Supplier No. MABF1105
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-DAG Kinase Alpha, clone 10D6.1 is a mouse monoclonal antibody that detects DAG kinase alpha. It has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAG Kinase Alpha
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10D6.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols DGKA,DAGK,DAGK1
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to the first 110 amino acids of human DAG kinase alpha.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001336
Test Specificity Clone 10D6.1 detects DAG kinase alpha in human. It targets an epitope within the first 110 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.