Learn More
Invitrogen™ DAGLA Polyclonal Antibody

Description
This antibody is predicted to react with rat based on sequence homology.
Specifications
Specifications
| Antigen | DAGLA |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | DAGLA |
| Gene Accession No. | Q6WQJ1, Q9Y4D2 |
| Gene Alias | C11orf11; DAGL(ALPHA); DAGLA; DAGLALPHA; DGL-alpha; diacylglycerol lipase alpha; diacylglycerol lipase, alpha; KIAA0659; Neural stem cell-derived dendrite regulator; NSDDR; Sn1-specific diacylglycerol lipase alpha |
| Gene Symbols | DAGLA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.