Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAGLA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564029
Description
Immunogen sequence: QEEPTYFAIW GDNKAFNEVI ISPAMLHEHL PYVVMEGLNK VLENYNKGKT ALLSAAKVMV SPTEVDLTPE LIFQQQPL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a diacylglycerol lipase. The encoded enzyme is involved in the biosynthesis of the endocannabinoid 2-arachidonoyl-glycerol.
Specifications
DAGLA | |
Polyclonal | |
Unconjugated | |
DAGLA | |
C11orf11; DAGL(ALPHA); DAGLA; DAGLALPHA; DGL-alpha; diacylglycerol lipase alpha; diacylglycerol lipase, alpha; KIAA0659; Neural stem cell-derived dendrite regulator; NSDDR; Sn1-specific diacylglycerol lipase alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
269060, 747 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6WQJ1, Q9Y4D2 | |
DAGLA | |
Recombinant protein corresponding to Human DAGLA. Recombinant protein control fragment (Product #RP-101750). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction