Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAGLB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567047 Supplier Invitrogen™ Supplier No. PA567047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVALDHRKESV VVAVRGTMSL QDVLTDLSAE SEVLDVECEV QDRLAHKGIS QAARYVYQRL INDGILSQAF SIA Highest antigen sequence identity to the following orthologs - mouse 78%, rat 80%.

Lipase that catalyzes the hydrolysis of arachidonic acid (AA)-esterified diacylglycerols (DAGs) to produce the principal endocannabinoid, 2-arachidonoylglycerol (2-AG) which can be further cleaved by downstream enzymes to release arachidonic acid (AA) for cyclooxygenase (COX)-mediated eicosanoid production (PubMed:14610053). Preferentially hydrolyzes DAGs at the sn-1 position in a calcium-dependent manner and has negligible activity against other lipids including monoacylglycerols and phospholipids (PubMed:14610053). Plays a key role in the regulation of 2-AG and AA pools utilized by COX1/2 to generate lipid mediators of macrophage and microglia inflammatory responses. Functions also as a polyunsaturated fatty acids-specific triacylglycerol lipase in macrophages. Plays an important role to support the metabolic and signaling demands of macrophages. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAGLB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DAGLB
Gene Accession No. Q8NCG7
Gene Alias DAGLB; DAGLBETA; DGL-beta; diacylglycerol lipase beta; diacylglycerol lipase, beta; E330036I19Rik; KCCR13L; RGD1310193; sn1-specific diacylglycerol lipase beta
Gene Symbols DAGLB
Host Species Rabbit
Immunogen Recombinant Human DAGLB. Recombinant protein control fragment (Product #RP-105476).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221955
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.