Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daidzin, 98%

Catalog No. p-7077838
Click to view available options
Quantity:
100 mg
1 g

CAS: 552-66-9 | C21H20O9 | 416.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-66-9
Molecular Formula C21H20O9
Molecular Weight (g/mol) 416.38
MDL Number MFCD00017466
InChI Key KYQZWONCHDNPDP-QNDFHXLGSA-N
Synonym daidzin, daidzoside, daidzein 7-o-glucoside, daidzein 7-glucoside, daidzein-7-glucoside, daidzein 7-o-beta-d-glucoside, unii-4r2x91a5m5, daidzein-7-o-glucoside, 7-beta-d-glucopyranosyloxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 7-o-b-d-glucopyranoside
PubChem CID 107971
ChEBI CHEBI:42202
IUPAC Name 3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES OC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Loss on Drying 2.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Formula Weight 416.38
Percent Purity 98%
Chemical Name or Material Daidzin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.