Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115091 Supplier Invitrogen™ Supplier No. PA5115091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DAK.

This gene is a member of the family of dihydroxyacetone kinases, which have a protein structure distinct from other kinases. The product of this gene phosphorylates dihydroxyacetone, and also catalyzes the formation of riboflavin 4',5'-phosphate (aka cyclin FMN) from FAD. Several alternatively spliced transcript variants have been identified, but the full-length nature of only one has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TKFC
Gene Accession No. Q3LXA3, Q4KLZ6, Q8VC30
Gene Alias ATP-dependent dihydroxyacetone kinase; BC021917; bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing); Dak; DHA kinase; Dha kinase/FMN cyclase; dihydroxyacetone kinase 2 homolog; dihydroxyacetone kinase 2 homolog (S. cerevisiae); DKFZP586B1621; DKFZP586B1621 protein; FAD-AMP lyase (cyclic FMN forming); FAD-AMP lyase (cyclizing); FAD-AMP lyase cyclic FMN forming; FAD-AMP lyase cyclizing; FMN cyclase; Glycerone kinase; MGC5621; NET45; RGD1311026; testis tissue sperm-binding protein Li 84P; TKFC; Triokinase; triokinase and FMN cyclase; triokinase, FMN cyclase; triokinase/FMN cyclase; Triose kinase; zgc:153296
Gene Symbols TKFC
Host Species Rabbit
Immunogen A synthesized peptide derived from human TKFC(Accession Q3LXA3), corresponding to amino acid residues G119-E169.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225913, 26007, 361730
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.