Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DALRD3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA557850

Catalog No. PIPA557850


Only null left
Add to Cart

Description

Description

Immunogen sequence: LQEALWHWPE DSHPGLAGAS DTGTGGCLVV HVVSCEEEFQ QQKLDLLWRK LVDKAPLRQK HLICGPVKVA GAPGTLMTAP EYYEFR Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.

The exact function of this gene is not known. It encodes a protein with a DALR anticodon binding domain similar to that of class Ia aminoacyl tRNA synthetases. This gene is located in a cluster of genes (with a complex sense-anti-sense genome architecture) on chromosome 3, and contains two micro RNA (miRNA) precursors (mir-425 and mir-191) in one of its introns. Preferential expression of this gene (the miRNAs and other genes in the cluster) in testis suggests a role of this gene in spermatogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DALRD3
Polyclonal
Unconjugated
DALRD3
DALR anticodon binding domain containing 3; DALR anticodon-binding domain-containing protein 3; DALRD3
Rabbit
Antigen Affinity Chromatography
RUO
55152
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5D0E6
DALRD3
Recombinant protein corresponding to Human DALRD3. Recombinant protein control fragment (Product #RP-96496).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.