Learn More
Invitrogen™ DAND5 Monoclonal Antibody (OTI2H3)

Description
DAND5 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | DAND5 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2H3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | DAND5 |
| Gene Accession No. | Q8N907 |
| Gene Alias | CER2; CER2CRL2; cerberus 2; Cerberus like-2; cerberus-like 2; cerberus-like protein 2; Cerl2; Cerl-2; Cerr2; CKTSF1B3; CKTSF1B3MGC126849; coco; CRL2; Cysteine knot superfamily 1, BMP antagonist 3; DAN domain BMP antagonist family member 5; DAN domain family member 5; DAN domain family member 5, BMP antagonist; DAN domain family, member 5; Dand5; DANTE; Dte; FLJ38607; GREM3; GREM3cerberus 2; gremlin-3; MGC126849; SP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.