Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dantrolene Sodium, USP, 90-96%, Spectrum™ Chemical
SDP

Catalog No. 18602016 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle

D3976, 24868-20-0, C14H9N4NaO5·3.5H2O

Spectrum™ Chemical Dantrolene Sodium, USP

Chemical Identifiers

CAS 24868-20-0
Molecular Formula C28H32N8Na2O17
Molecular Weight (g/mol) 798.58
InChI Key LTWQNYPDAUSXBC-CDJGKPBYSA-L
IUPAC Name disodium bis(3-[(E)-{[5-(4-nitrophenyl)furan-2-yl]methylidene}amino]-2,5-dioxoimidazolidin-1-ide) heptahydrate
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)\C=N\N1CC(=O)[N-]C1=O.[O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)\C=N\N1CC(=O)[N-]C1=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 90 to 96%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.