Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dantrolene Sodium, USP, 90-96%, Spectrum™ Chemical
SDP

Catalog No. 18602017 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-017 100 g Amber Glass Bottle
18-602-016 25 g Amber Glass Bottle
18-602-018 500 g Amber Glass Bottle
18-602-019 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-017 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D3976100GMBL
Add to Cart
Add to Cart

D3976, 24868-20-0, C14H9N4NaO5·3.5H2O

Spectrum™ Chemical Dantrolene Sodium, USP

Chemical Identifiers

CAS 24868-20-0
Molecular Formula C28H32N8Na2O17
Molecular Weight (g/mol) 798.58
InChI Key LTWQNYPDAUSXBC-CDJGKPBYSA-L
IUPAC Name disodium bis(3-[(E)-{[5-(4-nitrophenyl)furan-2-yl]methylidene}amino]-2,5-dioxoimidazolidin-1-ide) heptahydrate
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)\C=N\N1CC(=O)[N-]C1=O.[O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)\C=N\N1CC(=O)[N-]C1=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 90 to 96%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.