Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dantrolene Sodium, USP, 90-96%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Dantrolene Sodium, USP
Specifications
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 500 g |
Percent Purity | 90 to 96% |
Grade | USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction