Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAO Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297254MFV610
Description
DAO Polyclonal antibody specifically detects DAO in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
DAO | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
DAAO, D-amino-acid oxidase, DAMOXOXDA, DAO, EC 1.4.3, EC 1.4.3.3, MGC35381 | |
Produced in rabbits immunized with E. coli-derived Human DAO fragment. | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
1610 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction