Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597652
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597652 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597652 Supplier Invitrogen™ Supplier No. PA597652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAO Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes the peroxisomal enzyme D-amino acid oxidase. The enzyme is a flavoprotein which uses flavin adenine dinucleotide (FAD) as its prosthetic group. Its substrates include a wide variety of D-amino acids, but it is inactive on the naturally occurring L-amino acids. Its biological function is not known; it may act as a detoxifying agent which removes D-amino acids that accumulate during aging. In mice, it degrades D-serine, a co-agonist of the NMDA receptor. This gene may play a role in the pathophysiology of schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAO
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DAO
Gene Accession No. P14920
Gene Alias ABP; Abp1; AI987963; AOC1; DAAO; D-amino acid oxidase; D-amino acid oxidase (EC 1.4.3.3); D-amino acid oxidase 1; D-amino-acid oxidase; DAMOX; Dao; DAO1; Dao-1; Diamine oxidase; Histaminase; KAO; OXDA
Gene Symbols DAO
Host Species Rabbit
Immunogen Recombinant Human D-amino-acid oxidase protein (1-347AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1610
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.