Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP-5/p97 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982930
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2930 Supplier Bioss Supplier No. BS1350R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer′s disease. Alternative splicing results in multiple transcript variants.

Specifications

Antigen DAP-5/p97
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Eif4g2
Gene Accession No. P78344
Gene Alias AA589388; AAG1; aging-associated protein 1; DAP5; DAP-5; Death-associated protein 5; E130105L11Rik; eIF4G 2; eIF-4G 2; EIF4G2; eIF-4-gamma 2; Eukaryotic translation initiation factor 4 gamma 2; eukaryotic translation initiation factor 4 gamma, 2; eukaryotic translation initiation factor 4, gamma 2; eukaryotic translation initiation factor 4G-like 1; FLJ41344; Nat1; Natm1; Novel APOBEC-1 target 1; OK/SW-cl.75; p97; Translation repressor NAT1
Gene Symbols Eif4g2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DAP-5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1982
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.