Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP Kinase 1 Rabbit anti-Human, Mouse, Clone: 3E10G3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | DAP Kinase 1 |
---|---|
Clone | 3E10G3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
DAP Kinase 1 Monoclonal antibody specifically detects DAP Kinase 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
DAP Kinase 1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1612 | |
IgG | |
Affinity purified |
3E10G3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
DAPKDAP kinase 1, death-associated protein kinase 1, DKFZp781I035, EC 2.7.11, EC 2.7.11.1 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human DAP Kinase 1 (P53355). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title