Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP Kinase 2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen DAP Kinase 2
Concentration 0.5 mg/mL
Dilution Western Blot 2-4 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056344T
SDP
View Documents
Novus Biologicals
NB10056344SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056344
SDP
View Documents
Novus Biologicals
NB10056344
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

DAP Kinase 2 Polyclonal specifically detects DAP Kinase 2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

DAP Kinase 2
Western Blot 2-4 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat
Q9UIK4
23604
This antibody was developed against KLH-conjugated synthetic peptides corresponding to amino acids 325-342 and 347-364 of human DAPK2. It will cross-react with mouse and rat DAPK2.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Apoptosis
PBS containing 0.05% BSA with 0.05% Sodium Azide
DAP kinase 2, DAP-kinase-related protein 1, DAP-kinase-related protein 1 beta isoform, death-associated protein kinase 2, DRP1, DRP-1MGC119312, EC 2.7.11, EC 2.7.11.1
DAPK2
IgG
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.