Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP Kinase 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177291
Description
DAP Kinase 2 Polyclonal specifically detects DAP Kinase 2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DAP Kinase 2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
DAP kinase 2, DAP-kinase-related protein 1, DAP-kinase-related protein 1 beta isoform, death-associated protein kinase 2, DRP1, DRP-1MGC119312, EC 2.7.11, EC 2.7.11.1 | |
Rabbit | |
42 kDa | |
0.1 mg | |
Apoptosis, Autophagy, Cellular Markers | |
23604 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL | |
BAA88063 | |
DAPK2 | |
Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human DAPK2. This antigenic peptide is identical to the corresponding amino acids of mouse origin (1,2). The immunogen is located within the last 50 amino acids of DAPK2. Amino Acid Squence: ARRKALHPRRRSSTS | |
Affinity Purified | |
RUO | |
Primary | |
DAPK2 has no cross responses to DAPK1. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction