Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP Kinase 2 Antibody (OTI1C5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271753PCP
Description
DAP Kinase 2 Monoclonal specifically detects DAP Kinase 2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.Specifications
| DAP Kinase 2 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| DAPK2 | |
| Full length recombinant protein of human DAP Kinase 2 (NP_055141) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG3 |
| Flow Cytometry | |
| OTI1C5 | |
| Flow Cytometry | |
| DAP kinase 2, DAP-kinase-related protein 1, DAP-kinase-related protein 1 beta isoform, death-associated protein kinase 2, DRP1, DRP-1MGC119312, EC 2.7.11, EC 2.7.11.1 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Autophagy, Cellular Markers | |
| 23604.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction