Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534564
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534564 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534564 Supplier Invitrogen™ Supplier No. PA534564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human small intestine lysate. PA5-34564 can be used with blocking peptide PEP-1607.

This gene encodes a basic, proline-rich, 15-kD protein. The protein acts as a positive mediator of programmed cell death that is induced by interferon-gamma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DAP
Gene Accession No. P51397, Q91XC8, Q9QX67
Gene Alias 4921531N22Rik; AI789753; amylin peptide; AU040360; DAP; DAP 1; DAP1; DAP-1; death associated protein; death-associated protein; death-associated protein 1; MGC99796; OTTHUMP00000161670; OTTHUMP00000221331; Rap7a
Gene Symbols DAP
Host Species Rabbit
Immunogen A 19 amino acid peptide near the carboxy terminus of human DAP.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1611, 223453, 64322
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.