Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583090 Supplier Invitrogen™ Supplier No. PA583090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFRPNPTLLG NLRQNQVLCP LPYISIFKTR NSSSPNLVYG ESGWMSFEDH CAPRGAISRI CQDPRKILAL VLFQQSP Highest antigen sequence indentity to the following orthologs: Mouse - 27%, Rat - 25%.

This gene encodes a basic, proline-rich, 15-kD protein. The protein acts as a positive mediator of programmed cell death that is induced by interferon-gamma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DAP
Gene Accession No. P51397
Gene Alias 4921531N22Rik; AI789753; amylin peptide; AU040360; DAP; DAP 1; DAP1; DAP-1; death associated protein; death-associated protein; death-associated protein 1; MGC99796; OTTHUMP00000161670; OTTHUMP00000221331; Rap7a
Gene Symbols DAP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DAP. Recombinant protein control fragment (Product #RP-92025).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1611
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.