Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222259B
Description
DAP1 Polyclonal antibody specifically detects DAP1 in Human samples. It is validated for Western BlotSpecifications
DAP1 | |
Polyclonal | |
Western Blot | |
DAP1, DAP-1, death-associated protein, death-associated protein 1, MGC99796 | |
The immunogen this antibody was made to, maps to a region between residue 52 and 102 of human Death-Associated Protein using the numbering given in entry NP_004385.1 (GeneID 1611). | |
0.1 mL | |
Apoptosis, Cellular Markers | |
1611 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction