Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117352
Description
Centrifuge briefly prior to opening.
This gene encodes a basic, proline-rich, 15-kD protein. The protein acts as a positive mediator of programmed cell death that is induced by interferon-gamma.
Specifications
DAP1 | |
Polyclonal | |
Unconjugated | |
DAP | |
4921531N22Rik; AI789753; amylin peptide; AU040360; DAP; DAP 1; DAP1; DAP-1; death associated protein; death-associated protein; death-associated protein 1; MGC99796; OTTHUMP00000161670; OTTHUMP00000221331; Rap7a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1611 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.51 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P51397 | |
DAP | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human DAP1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction