Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987215
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7215 Supplier Bioss Supplier No. BS6351R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a basic, proline-rich, 15-kD protein. The protein acts as a positive mediator of programmed cell death that is induced by interferon-gamma.

Specifications

Antigen DAP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DAP
Gene Accession No. P51397
Gene Alias 4921531N22Rik; AI789753; AU040360; Dap; DAP 1; DAP1; DAP-1; death associated protein; death-associated protein; Death-associated protein 1; MGC99796; OTTHUMP00000161670; OTTHUMP00000221331; Rap7a
Gene Symbols DAP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DAP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1611
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.