Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP10/HCST Antibody (982107), DyLight 405, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB97862V
Description
DAP10/HCST Monoclonal specifically detects DAP10/HCST in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DAP10/HCST | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HCST | |
Synthetic peptide containing human DAP10/HCST | |
0.1 mL | |
Adaptive Immunity, Immune System Diseases, Immunology, Innate Immunity | |
Human | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
982107 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DAP10PIK3AP, DKFZP586C1522, hematopoietic cell signal transducer, KAP10DNAX-activation protein 10, kinase assoc pro of ∽10kDa, kinase assoc protein, Membrane protein DAP10, phosphoinositide-3-kinase adaptor protein, Transmembrane adapter protein KAP10 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction