Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP12 Mouse anti-Human, Alexa Fluor 594, Clone: 406209, R&D Systems™

Catalog No. FAB52401T10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB52401T10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB52401T10 Supplier R&D Systems Supplier No. FAB52401T100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DAP12 Monoclonal antibody specifically detects DAP12 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen DAP12
Applications Flow Cytometry
Classification Monoclonal
Clone 406209
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O43914
Gene Alias DAP12
Host Species Mouse
Immunogen Human Dap12 synthetic peptide, Accession # O43914
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.