Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP3 Antibody (42C617.1.2) - Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233222
Description
DAP3 Monoclonal specifically detects DAP3 in Human, Mouse samples. It is validated for Western Blot.Specifications
DAP3 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 3-5 ug/ml | |
bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
In Jurkat, a 45 kDa band is observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
42C617.1.2 | |
Unconjugated | |
P51398 | |
DAP3 | |
The antibody was raised against a synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein. | |
0.1 mg | |
Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
7818 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction