Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP3 Antibody (42C617.1.2), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056539MFV450
Description
DAP3 Monoclonal antibody specifically detects DAP3 in Human, Mouse samples. It is validated for Western BlotSpecifications
DAP3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
42C617.1.2 | |
Western Blot | |
bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
The antibody was raised against a synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein. | |
0.1 mL | |
Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
7818 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction