Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298784JF549
Description
DAP3 Polyclonal antibody specifically detects DAP3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
DAP3 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DAP3. | |
0.1 mL | |
Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
7818 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction