Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen DAP3
Concentration 0.5 mg/mL
Dilution Western Blot 1-2 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056325T
SDP
View Documents
Novus Biologicals
NB10056325SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056325
SDP
View Documents
Novus Biologicals
NB10056325
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

DAP3 Polyclonal specifically detects DAP3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

DAP3
Western Blot 1-2 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat
P51398
7818
The antibody was raised against a synthetic peptide corresponding to amino acids 44-57 (TSDSDPAKHGEQHE) of mouse DAP-3 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Apoptosis, Core ESC Like Genes, Stem Cell Markers
PBS with 0.02% Sodium Azide
bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt
DAP3
IgG
Protein G purified
In Jurkat, a 45 kDa band is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.