Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP3 Antibody (OTI1B12), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271338AF750
Description
DAP3 Monoclonal specifically detects DAP3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DAP3 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
DAP3 | |
Recombinant protein fragment corresponding to amino acids 161-398 of human DAP3 (NP_387506) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1B12 | |
Western Blot, Immunohistochemistry | |
bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
Mouse | |
Affinity Purified | |
Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
7818.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction