Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAP3 Monoclonal Antibody (42C617.1.2)
GREENER_CHOICE

Catalog No. 01676609
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-609 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-609 Supplier Invitrogen™ Supplier No. MA141279
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This sequence is 75% homologous in mouse and rat. Suggested positive control: Jurkat, antigen standard for DAP3 (transient overexpression lysate), Jurkat whole cell lysate.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that also participates in apoptotic pathways which are initiated by tumor necrosis factor-alpha, Fas ligand, and gamma interferon. This protein potentially binds ATP/GTP and might be a functional partner of the mitoribosomal protein S27. Splice variants that differ in the 5' UTR have been found for this gene; both variants encode the same protein. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAP3
Applications Western Blot
Classification Monoclonal
Clone 42C617.1.2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene DAP3
Gene Accession No. P51398, Q9ER88
Gene Alias 28S ribosomal protein S29, mitochondrial; 4921514D13Rik; bMRP-10; DAP3; DAP-3; death associated protein 3; death-associated protein 3; DKFZp686G12159; Ionizing radiation resistance conferring protein; MGC126058; MGC126059; mitochondrial 28S ribosomal protein S29; Mitochondrial small ribosomal subunit protein mS29; MRPS29; MRP-S29; S29mt
Gene Symbols DAP3
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65111, 7818
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.