Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596541 Supplier Invitrogen™ Supplier No. PA596541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLWVKNCRDL LQSSYNKQRF DQPLEASTWL KNFKTTNERF LNQIKVQEKY VWNKRESTEK GSPLGEVVEQ GITRVRNATD AVGIVLKELK RQSSLGMFHL LVAVDGINAL WGRTTLKRED KSPIAPEELA LVHNLRKMMK NDWHGGAIVS ALSQTGSLFK PRKAYLPQEL LGKEGFDALD PFIPILVSNY NPKEFESCIQ YYLENNWLQH EKAPTEEGKK ELLFLSNANP SLLERHCAYL; Positive Samples: MCF7, Jurkat, Mouse kidney, Mouse liver; Cellular Location: Mitochondrion.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that also participates in apoptotic pathways which are initiated by tumor necrosis factor-alpha, Fas ligand, and gamma interferon. This protein potentially binds ATP/GTP and might be a functional partner of the mitoribosomal protein S27. Splice variants that differ in the 5' UTR have been found for this gene; both variants encode the same protein. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DAP3
Gene Accession No. P51398, Q9ER88
Gene Alias 28S ribosomal protein S29, mitochondrial; 4921514D13Rik; bMRP-10; DAP3; DAP-3; death associated protein 3; death-associated protein 3; DKFZp686G12159; Ionizing radiation resistance conferring protein; MGC126058; MGC126059; mitochondrial 28S ribosomal protein S29; Mitochondrial small ribosomal subunit protein mS29; MRPS29; MRP-S29; S29mt
Gene Symbols DAP3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 159-398 of human DAP3 (NP_3875061).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295238, 65111, 7818
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.