Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA141658
Description
Suggested positive control: Jurkat, antigen standard for DAP3 (transient overexpression lysate), Jurkat whole cell lysate. Immunogen sequence: TSDSDPAKHG EQHE.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that also participates in apoptotic pathways which are initiated by tumor necrosis factor-alpha, Fas ligand, and gamma interferon. This protein potentially binds ATP/GTP and might be a functional partner of the mitoribosomal protein S27. Splice variants that differ in the 5' UTR have been found for this gene; both variants encode the same protein. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
Specifications
| DAP3 | |
| Polyclonal | |
| Unconjugated | |
| DAP3 | |
| 28S ribosomal protein S29, mitochondrial; 4921514D13Rik; bMRP-10; DAP3; DAP-3; death associated protein 3; death-associated protein 3; DKFZp686G12159; Ionizing radiation resistance conferring protein; MGC126058; MGC126059; mitochondrial 28S ribosomal protein S29; Mitochondrial small ribosomal subunit protein mS29; MRPS29; MRP-S29; S29mt | |
| Rabbit | |
| Protein G | |
| RUO | |
| 295238, 65111, 7818 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.02% sodium azide | |
| P51398, Q9ER88 | |
| DAP3 | |
| Synthetic peptide corresponding to amino acids 44-57 of mouse DAP-3 protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction