Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586453
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586453 Supplier Invitrogen™ Supplier No. PA586453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer's disease. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAP5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Eif4g2
Gene Accession No. P78344, Q62448
Gene Alias AA589388; AAG1; aging-associated protein 1; DAP5; DAP-5; Death-associated protein 5; E130105L11Rik; eIF4G 2; eIF-4G 2; EIF4G2; eif4g2.L; eif4g2.S; eIF-4-gamma 2; Eukaryotic translation initiation factor 4 gamma 2; eukaryotic translation initiation factor 4 gamma, 2; eukaryotic translation initiation factor 4 gamma, 2 L homeolog; eukaryotic translation initiation factor 4 gamma, 2 S homeolog; eukaryotic translation initiation factor 4 gamma, 2 S homeolog; eukaryotic translation initiation factor 4, gamma 2; eukaryotic translation initiation factor 4G-like 1; FLJ41344; NAT1; Natm1; Novel APOBEC-1 target 1; OK/SW-cl.75; p97; translation repressor NAT1; XELAEV_18024574mg
Gene Symbols Eif4g2
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 35-80 of Human DAP-5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13690, 1982, 361628
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.